Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylphosphonic acid, 98%, Thermo Scientific Chemicals

Catalog No. p-7026747
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2293922 100 g
AAB2293936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2293922 Supplier Thermo Scientific Chemicals Supplier No. B2293922
Only null left
Add to Cart

Phenylphosphonic acid additive used with Noyori's catalyst for oxidation of sulfides to sulfones. Used as additive of unsaturated polyester PU resin. It also can be added into nylon to improve its polymerization degree. It can be used as extreme pressure agent of lube; used as catalyst of carboxylic acid & alcohol reaction; it can be used as fire retarding treatment of fiber used asganic materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenylphosphonic acid additive used with Noyori′s catalyst for oxidation of sulfides to sulfones. Used as additive of unsaturated polyester PU resin. It also can be added into nylon to improve its polymerization degree. It can be used as extreme pressure agent of lube; used as catalyst of carboxylic acid & alcohol reaction; it can be used as fire retarding treatment of fiber used asganic materials.

Solubility
Soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1571-33-1
Molecular Formula C6H7O3P
Molecular Weight (g/mol) 158.09
MDL Number MFCD00002136
InChI Key QLZHNIAADXEJJP-UHFFFAOYSA-N
Synonym benzenephosphonic acid, phosphonic acid, phenyl, phenyl-phosphonic acid, phosphonic acid, p-phenyl, phenyl phosphonic acid, unii-byd76t2868, sv7, pubchem21303, phosphonic acid,phenyl, wln: qpqo&r
PubChem CID 15295
IUPAC Name phenylphosphonic acid
SMILES OP(O)(=O)C1=CC=CC=C1

Specifications

Melting Point 160°C to 162°C
Quantity 100 g
UN Number UN3261
Beilstein 2245168
Solubility Information Soluble in water.
Formula Weight 158.09
Percent Purity 98%
Chemical Name or Material Phenylphosphonic acid
Hazard Category H302-H315-H318-H335
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 216-388-1
RTECSNumber TA0350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.