Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylselenol, 90+%

Catalog No. AAA1287606
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1287606 5 g
AAA1287603 1 g
AAA1287614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1287606 Supplier Thermo Scientific Chemicals Supplier No. A1287606
Add to Cart

CAS: 645-96-5 | C6H6Se

It is applied for reactions of phenylselenol and its anion. Benzeneselenol can used as starting reagent in the synthesis of monoseleno-substituted 1,3-dienes. It may be used in the synthesis of cationic chalcogenolato-bridged diruthenium complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied for reactions of phenylselenol and its anion. Benzeneselenol can used as starting reagent in the synthesis of monoseleno-substituted 1,3-dienes. It may be used in the synthesis of cationic chalcogenolato-bridged diruthenium complexes.

Solubility
Not miscible or difficult to mix in water.

Notes
Air sensitive. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 645-96-5
Molecular Formula C6H6Se
MDL Number MFCD00000001
Synonym Benzeneselenol; Selenophenol

Specifications

Density 1.479
Boiling Point 71°C to 72°C (18 mmHg)
Flash Point 70°C (158°F)
Odor Stench
Refractive Index 1.612
Quantity 5 g
UN Number UN3440
Beilstein 385715
Sensitivity Air sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 157.07
Percent Purity ≥90%
Chemical Name or Material Phenylselenol
Show More Show Less
Hazard Category H227-H301+H331-H373
Hazard Statement GHS H Statement
H301-H331-H373-H227
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P301+P310-P304+P340-P311-P314-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: SELENIUM COMPOUND, LIQUID, N.O.S.
EINECSNumber 211-457-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.