Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylsulfonylacetonitrile, 98%, Thermo Scientific™

Catalog No. AAA1542730
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1542730 250 g
AAA1542718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1542730 Supplier Thermo Scientific Chemicals Supplier No. A1542730

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7605-28-9
Molecular Formula C8H7NO2S
Molecular Weight (g/mol) 181.209
MDL Number MFCD00007550
InChI Key ZFCFFNGBCVAUDE-UHFFFAOYSA-N
Synonym benzenesulfonylacetonitrile, phenylsulfonyl acetonitrile, 2-phenylsulfonyl acetonitrile, phenylsulfonylacetonitrile, 2-benzenesulfonyl acetonitrile, phenylsulphonylacetonitrile, acetonitrile, phenylsulfonyl, benzenesulphonyl acetonitrile, benzenesulfonyl-acetonitrile, 2-phenylsulfonyl ethanenitrile
PubChem CID 82077
IUPAC Name 2-(benzenesulfonyl)acetonitrile
SMILES C1=CC=C(C=C1)S(=O)(=O)CC#N

Specifications

Melting Point 110°C to 114°C
Quantity 250 g
UN Number UN3439
Beilstein 640716
Formula Weight 181.22
Percent Purity 98%
Chemical Name or Material Phenylsulfonylacetonitrile
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 231-515-0
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.