Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Phenylthio)acetic Acid 98.0+%, TCI America™
SDP

Catalog No. P0753250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0753250G 250 g
P075325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0753250G Supplier TCI America Supplier No. P0753250G

Chemical Identifiers

CAS 103-04-8
Molecular Formula C8H7O2S
Molecular Weight (g/mol) 167.20
MDL Number MFCD00004355
InChI Key MOTOSAGBNXXRRE-UHFFFAOYSA-M
Synonym phenylthio acetic acid, thiophenoxyacetic acid, 2-phenylthio acetic acid, phenylthioglycolic acid, phenylmercaptoacetic acid, 2-phenylsulfanyl acetic acid, phenylsulfanyl acetic acid, acetic acid, phenylthio, phenylmercapto acetic acid, carboxymethyl phenyl sulfide
PubChem CID 59541
IUPAC Name 2-(phenylsulfanyl)acetate
SMILES [O-]C(=O)CSC1=CC=CC=C1

Specifications

Melting Point 64°C
Color White-Yellow
Quantity 250 g
Formula Weight 168.21
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (Phenylthio)acetic Acid
EINECSNumber (3)-1103
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.