Learn More
Sigma Organic Chemistry Phenylthiohydantoin-glu | 10mg | MFCD00038436
Supplier: Sigma Organic Chemistry CDS01326610MG

Phenylthiohydantoin-glu , 10mg
About this Item:
MDL #: MFCD00038436
Molecular Weight: 263.32
UNSPSC Code: 12352200
Chemical Formula: C12H13N3O2S
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.