Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylthiohydantoin-proline 98.0+%, TCI America™
SDP

Catalog No. P0729100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0729100MG 100 mg
P07291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0729100MG Supplier TCI America Supplier No. P0729100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4333-21-5
Molecular Formula C12H12N2OS
Molecular Weight (g/mol) 232.301
MDL Number MFCD00056895
InChI Key YNJJBMAKBUZCKN-UHFFFAOYSA-N
Synonym PTH-proline
PubChem CID 100798
IUPAC Name 2-phenyl-3-sulfanylidene-5,6,7,7a-tetrahydropyrrolo[1,2-c]imidazol-1-one
SMILES C1CC2C(=O)N(C(=S)N2C1)C3=CC=CC=C3

Specifications

Melting Point 179°C
Color White
Quantity 100 mg
Formula Weight 232.30
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Phenylthiohydantoin-proline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.