Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylurea 98.0+%, TCI America™
SDP

Catalog No. P0247100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0247100G 100 g
P024725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0247100G Supplier TCI America Supplier No. P0247100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 64-10-8
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.154
MDL Number MFCD00007944
InChI Key LUBJCRLGQSPQNN-UHFFFAOYSA-N
Synonym 1-phenylurea, n-phenylurea, urea, phenyl, phenylcarbamide, monophenylurea, stabilizer vh, stabilisator vh, phenyl-urea, urea, n-phenyl, phenyl urea
PubChem CID 6145
IUPAC Name phenylurea
SMILES C1=CC=C(C=C1)NC(=O)N

Specifications

Melting Point 147°C
Color White-Yellow
Boiling Point 238°C
Quantity 100 g
Formula Weight 136.15
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Phenylurea
EINECSNumber (9)-1662
RTECSNumber YU0650000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.