Learn More
Selleck Chemical LLC Clomipramine HCl S2541-1g
SDP
Supplier:  Selleck Chemical LLC S25411G
Encompass_Preferred
Clomipramine HCl is a tricyclic antidepressant acting as a potent inhibitor of 5-HT reuptake with an IC50 value of 1 5 nM Clomipramine hydrochloride is used in research on depression anxiety and obsessive-compulsive disorder (OCD)
Catalog No. 50-311-1638
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.