Learn More
Description
MIP0802 targets Phenytoin (Dilantin) in ELISA, IP, and RIA applications and shows reactivity with Chemical samples. The MIP0802 immunogen is phenytoin-KLH. MIP0802 was formerly sold as a Seradyn product.
Specifications
Specifications
| Antigen | Phenytoin |
| Applications | ELISA, Immunoprecipitation, Radioimmune Assays (RIA) |
| Classification | Monoclonal |
| Clone | 1P1H2 |
| Concentration | 5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene Alias | Dilantin; Epanutin; Infatabs; Kapseals; Phenytek |
| Host Species | Mouse |
| Immunogen | Phenytoin-KLH. |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
