Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phenytoin Monoclonal Antibody (PY25-500.1)
GREENER_CHOICE

Catalog No. 01671969 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-671-969 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-671-969 Supplier Invitrogen™ Supplier No. MA110739
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-10739 detects Phenytoin. MA1-10739 has been successfully used in ELISA applications. The MA1-10739 immunogen is Phenytoin, conjugated to KLH.

Phenytoin sodium is a commonly used antiepileptic. Phenytoin (fen-i-TOE-in) acts to suppress the abnormal brain activity seen in seizure by reducing electrical conductance among brain cells by stabilizing the inactive state of voltage-gated sodium channels. Aside from seizures, it is an option in the treatment of trigeminal neuralgia in the event that carbamazepine is deemed inappropriate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phenytoin
Applications ELISA
Classification Monoclonal
Clone PY25-500.1
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias Dilantin; Epanutin; Infatabs; Kapseals; Phenytek
Host Species Mouse
Immunogen Phenytoin, conjugated to KLH.
Purification Method Protein G
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.