Learn More
Apexbio Technology LLC Phenytoin sodium, 1g Cas# 630-93-3
SDP
Supplier:  Apexbio Technology LLC B22721000
Encompass_Preferred
Phenytoin is an inactive voltage-gated sodium channel stabilizer. 630-93-3. MFCD00069674 Phenytoin is an inactive voltage-gated sodium channel stabilizer.Phenytoin is an antiepileptic drug. It is useful to treat partial seizures and generalized tonic-clonic seizures but not primary generalized seizures such as absence seizures or myoclonic se There are other sizes available. Please inqury us for quote.
Catalog No. 50-199-8693
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.