Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597060
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597060 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597060 Supplier Invitrogen™ Supplier No. PA597060
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAQPPRLSRS GASSLWDPAS PAPTSGPRPR LWEGQDVLAR WTDGLLYLGT IKKVDSAREV CLVQFEDDSQ FLVLWKDISP AALPGEELLC CVCRSETVVP GNRLVSCEKC RHAYHQDCHV PRAPAPGEGE GTSWVCRQCV FAIATKRGGA LKKGPYARAM LGMKLSLPYG LKGLDWDAGH LSNRQQSYCY CGGPGEWNLK MLQCRSCLQW FHEACTQCLS KPLLYGDRFY EFECCVCRGG PEKVRRLQLR WVDVAHLVLY; Positive Samples: A-549, HeLa, Mouse lung; Cellular Location: Cytoplasm, Nucleus, centrosome, cytoskeleton, microtubule organizing center.

This gene encodes a protein with significant sequence similarity to Drosophila Polycomblike. The encoded protein contains a zinc finger-like PHD (plant homeodomain) finger which is distinct from other classes of zinc finger motifs and which shows the typical Cys4-His-Cys3 arrangement. PHD finger genes are thought to belong to a diverse group of transcriptional regulators possibly affecting eukaryotic gene expression by influencing chromatin structure. Two transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHF1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.87 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PHF1
Gene Accession No. O43189, Q9Z1B8
Gene Alias AW557215; D17Ertd455e; hPCl1; hPHF1; mPCl1; MTF2L2; PCL1; PHD finger protein 1; PHF1; Phf2; Plc1; polycomb-like 1; polycomb-like protein 1; Protein PHF1; T-complex testis-expressed 3; Tctex3; Tctex-3; TDRD19C; testicular tissue protein Li 140; tudor domain containing 19C
Gene Symbols PHF1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human PHF1 (NP_0770841).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21652, 294287, 5252
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.