Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHF13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA524986
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA524986 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA524986 Supplier Invitrogen™ Supplier No. PA524986
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with mouse based on sequence homology.

PHF13 contains 1 PHD-type zinc finger. The function of PHF13 is currently unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHF13
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene PHF13
Gene Accession No. Q86YI8, Q8K2W6
Gene Alias BC029632; PHD finger protein 13; PHD zinc finger protein PHF5; PHF13; PHF5; SPOC1; Survival time-associated PHD finger protein in ovarian cancer 1; survival time-associated PHD protein in ovarian cancer
Gene Symbols PHF13
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 263-291 amino acids from the C-terminal region of human PHF13.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100760522, 148479, 230936
Target Species Human, Mouse, Hamster
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.