Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHF19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596915
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596915 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596915 Supplier Invitrogen™ Supplier No. PA596915
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Mouse brain, Rat brain; Cellular Location: Nucleus Immunogen sequence: MENRALDPGT RDSYGATSHL PNKGALAKVK NNFKDLMSKL TEGQYVLCRW TDGLYYLGKI KRVSSSKQSC LVTFEDNSKY WVLWKDIQHA GVPGEEPKCN ICLGKTSGPL NEILICGKCG LGYHQQCHIP IAGSADQPLL TPWFCRRCIF ALAVRVSLPS SPVPASPASS SGADQRLPSQ SLSSKQKGHT WALETDSASA TVLGQDL.

PHF19 contains 2 PHD-type zinc fingers. It acts as a transcriptional repressor. Isoform 1 and isoform 2 inhibit transcription from an HSV-tk promoter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHF19
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.63 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene PHF19
Gene Accession No. Q5T6S3, Q9CXG9
Gene Alias 3110009G19Rik; 3321402G02Rik; DKFZP727G051; hPCL3; MTF2L1; Pcl3; PHD finger protein 19; phf13; Phf19; polycomb like 3; polycomb-like 3; polycomb-like protein 3; si:dkeyp-51b7.1; TDRD19B; tudor domain containing 19B
Gene Symbols PHF19
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-207 of human PHF19 (NP_0010099361).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26147, 296653, 74016
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.