Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA14120
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA14120 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA14120 Supplier Invitrogen™ Supplier No. PA14120
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-4120 detects PHF-2 in human samples. PA1-4120 has been successfully used in Western blot procedures. By Western blot this antibody detects a ~120 kDa protein representing PHF-2 in human placenta lysate. The PA1-4120 immunogen is a synthetic peptide corresponding to residues K(70) H G P G P T P D V K R V C(82) of the 120 kDa PHF.

This gene encodes a protein which contains a zinc finger-like PHD (plant homeodomain) finger, distinct from other classes of zinc finger motifs, and a hydrophobic and highly conserved domain. The PHD finger shows the typical Cys4-His-Cys3 arrangement. PHD finger genes are thought to belong to a diverse group of transcriptional regulators possibly affecting eukaryotic gene expression by influencing chromatin structure.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHF2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene PHF2
Gene Accession No. O75151
Gene Alias CENP-35; centromere protein 35; GRC5; JHDM1E; jumonji C domain-containing histone demethylase 1E; Kiaa0662; Lysine-specific demethylase PHF2; PHD finger protein 2; Phf2
Gene Symbols PHF2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues K(70) H G P G P T P D V K R V C(82) of the 120 kDa PHF.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5253
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.