Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHF20 Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501993163
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3163 10 μL
50-199-3162 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3163 Supplier Active Motif Supplier No. 61124
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Methyllysine-binding protein, component of the MOF histone acetyltransferase protein complex. Not required for maintaining the global histone H4 'Lys-16' acetylation (H4K16ac) levels or locus specific histone acetylation, but instead works downstream in transcriptional regulation of MOF target genes. As part of the NSL complex it may be involved in acetylation of nucleosomal histone H4 on several lysine residues. Contributes to methyllysine-dependent p53/TP53 stabilization and up-regulation after DNA damage.

Specifications

Antigen PHF20
Applications Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
Gene PHF20
Gene Accession No. Q8BLG0, Q9BVI0
Gene Alias 6820402O20Rik; BOS_13202; C20orf104; GLEA2; Glioma-expressed antigen 2; Hca58; Hepatocellular carcinoma-associated antigen 58; Hepatocellular carcinoma-associated antigen 58 homolog; Novel zinc finger protein; NZF; PHD finger protein 20; Phf20; TDRD20A; Transcription factor TZP; tudor domain containing 20A; TZP
Gene Symbols PHF20
Host Species Rabbit
Immunogen This PHF20 antibody was raised against a peptide derived from human PHF20.
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 228829, 51230
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.