Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHF21B Monoclonal Antibody (OTI3G9), TrueMAB™, OriGene
SDP

Catalog No. p-200011905
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1136 30 μL
50-167-4044 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1136 Supplier OriGene Supplier No. TA503072S
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PHF21B Monoclonal antibody specifically detects PHF21B in Human samples. It is validated for Flow Cytometry, Western Blot

Specifications

Antigen PHF21B
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI3G9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene SH2B3
Gene Accession No. Q9UQQ2
Gene Alias A730032D07Rik; BHC80L; KIAA1661; PHD finger protein 21B; PHD finger protein 4; PHF21B; PHF4; RGD1308739
Host Species Mouse
Immunogen Full length human recombinant protein of human SH2B3 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10019
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.