Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHF23 Polyclonal Antibody, Bethyl Laboratories

Rabbit Polyclonal Antibody

$151.31

Specifications

Antigen PHF23
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Liquid
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-156-2305
Encompass_Preferred
View Documents
Bethyl Laboratories, Inc
A302320AT
20 μL
Each of 1 for $151.31
Only null left
Add to Cart
 
Description

Description

The recommended shelf life for this product is 1 year from date of receipt.

Acts as a negative regulator of autophagy, through promoting ubiquitination and degradation of LRSAM1, an E3 ubiquitin ligase that promotes autophagy in response to starvation or infecting bacteria.
Specifications

Specifications

PHF23
Polyclonal
Liquid
RUO
TBS with 0.1% BSA and 0.09% sodium azide
FLJ16355, hJUNE-1b, MGC2941, PDH-containing protein JUNE-1, PHD finger protein 23a
PHF23
IgG
Antigen affinity chromatography
Antibody
Western Blot
Unconjugated
Rabbit
Human
Q9BUL5
79142
Between 250 and 300
Primary
4° C
PHF23
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.