Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phi29 DNA Polymerase, 1250 U, G-Biosciences
SCI_ED

Catalog No. S001148
Change view
Click to view available options
Form:
Liquid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
S001148 Liquid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S001148 Supplier G-Biosciences Supplier No. 7862261
Add to Cart
Add to Cart

phi29 DNA Polymerase is a highly processive polymerase with strong strand displacement activity and a powerful 3' → 5' proofreading exonuclease activity.

  • It is used to synthesize new DNA strands by adding nucleotides to a template strand.

Specifications

Concentration 10 U/μL
Detection Method Gel Electrophoresis
Form Liquid
GC-Rich PCR Performance Medium
PCR Method End-point PCR
Reaction Speed Fast
Label or Dye FAM, Biotin
Product Type phi29 DNA Polymerase
For Use With (Application) DNA Sequencing, Isothermal Amplification
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.