Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Philanthotoxin 74, Tocris Bioscience™

Catalog No. 27701
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-701 1 mg
27-701-0 10 mg
27-705-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 27-701 Supplier R&D Systems Supplier No. 2770/1

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

GluA1 and GluA3 selective AMPA antagonist

AMPA receptor antagonist. Non-selective homomeric GluA3 and GluA1 receptor antagonist (IC50 values are 263 and 296 nM respectively). Inhibits homomeric GluA3 and GluA1 with a 100-fold greater potency than heteromeric GluA1/2 and GluA2/3 receptors.

Chemical Identifiers

CAS 1227301-51-0
Molecular Formula C24H44Cl2N4O3
Molecular Weight (g/mol) 507.541
InChI Key HWTJQQMIKVJWLH-IKXQUJFKSA-N
Synonym philanthotoxin 74, philanthotoxin 74 dihydrochloride, phtx-74, philanthotoxin-7,4, phtx-74 dihydrochloride, n-7-4-aminobutyl amino heptyl-nalpha-butanoyl-l-tyrosinamide-hydrogen chloride 1/2, s-n-7-4-aminobutyl amino heptyl-4-hydroxy-?-1-oxobutyl amino benzenepropanamide dihydrochloride, s-n-7-4-aminobutyl amino heptyl-4-hydroxy-alpha-1-oxobutyl amino benzenepropanamide dihydrochloride, n-1s-1-7-4-aminobutyl amino heptyl carbamoyl-2-4-hydroxyphenyl ethyl butanamide dihydrochloride
PubChem CID 46213501
IUPAC Name N-[(2S)-1-[7-(4-aminobutylamino)heptylamino]-3-(4-hydroxyphenyl)-1-oxopropan-2-yl]butanamide;dihydrochloride
SMILES CCCC(=O)NC(CC1=CC=C(C=C1)O)C(=O)NCCCCCCCNCCCCN.Cl.Cl

Specifications

Quantity 1 mg
Formula Weight 507.54
Percent Purity >98%
Chemical Name or Material Philanthotoxin 74
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.