Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHIP Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501733162 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-3162 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-3162 Supplier Proteintech Group Inc Supplier No. 209331AP150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

PHIP (pleckstrin homology domain interacting protein), also known as ndrp or WDR11, is a 1,821 amino acid protein that contains eight N-terminal WD40 repeats and two bromodomains. It is expressed in skeletal muscle (localizing to the cytosol and nucleus) and primary beta cells (localizing to the nucleus) and acts as a transcriptional activator. PHIP is known to interact with various members of the insulin receptor substrate (IRS) family. The IRS family of proteins mediate insulin receptor signaling and play an important role in insulin-producing beta cell proliferation and survival. PHIP specifically associates with the PH domain of IRS-1 and may function to link IRS-1 to insulin receptors, indicating a vital role of PHIP in the regulation of insulin signaling.

Specifications

Antigen PHIP
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.82 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PHIP
Gene Accession No. Q8VDD9, Q8WWQ0
Gene Alias ndrp, PH interacting protein, PHIP, WDR11
Gene Symbols PHIP
Host Species Rabbit
Immunogen PHIP Fusion Protein Ag14968
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55023, 83946
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.