Learn More
Santa Cruz Biotechnology PHISTONE H2A.X ANTBDY 100UG/UL
Supplier:  Santa Cruz Biotechnology SC517348
Encompass
p-Histone H2A.X Antibody (Ser 139) is an IgG1 mouse monoclonal p-Histone H2A.X antibody (also designated p-Histone H2A.X antibody or Histone H2A antibody) that detects Ser 139 phosphorylated Histone H2A.X of mouse, rat and human origin by WB, IP and IF. p-Histone H2A.X Antibody (Ser 139) is available as the non-conjugated anti-p-Histone H2A.X antibody form. Histone H2A.X is a member of the Histone H2A family, which is involved in nucleosomal organization of chromatin. The H2AFX gene is located in close proximity to the Porphobilinogen deaminase (PBG-D) gene in both mouse and human, and maps to chromosome 9 and 11q23, respectively. H2A.X differs from the other members of the H2A family by the presence of a highly conserved C-terminal motif. It is rapidly phosphorylated in response to ionizing radiation and plays an important role in the recognition and repair of DNA double stranded breaks.
Catalog No. NC2387148
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.