Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHKG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575548
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575548 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575548 Supplier Invitrogen™ Supplier No. PA575548
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

PHKG1 is a serine/threonine kinase with glycogenolytic regulatory functions. This protein is the catalytic member of a 16 subunit protein kinase complex which contains equimolar ratios of 4 subunit types. The complex is a crucial glycogenolytic regulatory enzyme. PHKG1 has two pseudogenes at chromosome 7q11.21 and one at chromosome 11p11.12.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHKG1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene PHKG1
Gene Accession No. P07934, P13286, Q16816
Gene Alias gamma phosphorylase kinase; MGC137450 protein; PHGK; PHKG; PHKG1; PHKgammaM; PHK-gammaM; PHK-gamma-M; PHKgamma-M; PHKIN01; phosphorylase b kinase gamma catalytic chain, skeletal muscle isoform; Phosphorylase B kinase gamma catalytic chain, skeletal muscle isoform (Phosphorylase kinase gamma subunit 1); phosphorylase b kinase gamma catalytic chain, skeletal muscle/heart isoform; phosphorylase kinase; phosphorylase kinase catalytic subunit gamma 1; phosphorylase kinase gamma; phosphorylase kinase gamma 1; phosphorylase kinase gamma subunit; phosphorylase kinase gamma subunit 1; phosphorylase kinase subunit gamma 1; phosphorylase kinase subunit gamma-1; phosphorylase kinase, gamma 1; phosphorylase kinase, gamma 1 (muscle); serine/threonine-protein kinase PHKG1; unnamed protein product
Gene Symbols PHKG1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 293-338 of Human PHKG1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18682, 29353, 5260
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.