Learn More
Sigma Organic Chemistry Phlorizin hydrate | 100mg | MFCD09037358
Supplier: Sigma Organic Chemistry CDS000104100MG

Phlorizin hydrate , 100mg
About this Item:
MDL #: MFCD09037358
Molecular Weight: 454.42
UNSPSC Code: 12352200
Chemical Formula: C21H26O11
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.