Description
- For the quantitative determination of sodium, potassium and chloride concentrations in serum and urine samples
- Solutions available include buffer, mid-standard, reference, low serum standard, high serum standard, low/high urine standard, internal reference fill, Na/K selectivity check, cleaning and wash
Specifications
Specifications
| Type | Mid Standard |
| For Use With (Equipment) | Olympus™ AU series analyzer |
| Capacity (Metric) | 4 x 2000 mL |
| Reagents | Mid Standard |