Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phorbol 12-Myristate 13-Acetate (Clear or Whitish Film), Fisher BioReagents™

Catalog No. BP6851 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
Packaging:
Glass Ampule
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP685-1 1 mg Glass Ampule
BP685-5 5 mg Glass Ampule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP685-1

Phorbol 12-Myristate 13-Acetate (Clear or Whitish Film)

PMA is an activator of Protein Kinase C; it is a skin irritant and mouse skin tumor promoter.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16561-29-8
Molecular Formula C36H56O8
Molecular Weight (g/mol) 616.84
MDL Number MFCD00036736
InChI Key PHEDXBVPIONUQT-RGYGYFBISA-N
Synonym phorbol 12-myristate 13-acetate, phorbol ester, 12-o-tetradecanoylphorbol-13-acetate, 12-o-tetradecanoylphorbol 13-acetate, factor a1, tetradecanoylphorbol acetate, 12-tetradecanoylphorbol 13-acetate, factor a1 croton oil, phorbol 13-acetate 12-myristate, phorbol-12-myristate-13-acetate
PubChem CID 27924
ChEBI CHEBI:37537
IUPAC Name (1S,2S,6R,10S,11R,13S,14R,15R)-13-(acetyloxy)-1,6-dihydroxy-8-(hydroxymethyl)-4,12,12,15-tetramethyl-5-oxotetracyclo[8.5.0.0²,⁶.0¹¹,¹³]pentadeca-3,8-dien-14-yl tetradecanoate
SMILES CCCCCCCCCCCCCC(=O)O[C@@H]1[C@@H](C)[C@]2(O)[C@@H]3C=C(C)C(=O)[C@@]3(O)CC(CO)=C[C@H]2[C@@H]2C(C)(C)[C@]12OC(C)=O

Specifications

Melting Point 72°C
Color White
Assay Percent Range ≥99.0 %
Packaging Glass Ampule
Quantity 1 mg
Formula Weight 616.8
Percent Purity 99.0% min.
Physical Form Solid
Chemical Name or Material Phorbol 12-Myristate 13-Acetate
Safety handling Check Alert storage code Red
Hazard Category Signal Word: WARNING!
Precautionary Statement Emergency Overview
Irritating to eyes, respiratory system and skin.
NFPA
Health: 1
Flammability:
Recommended Storage -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.