CAS RN 16561-29-8
Phorbol 12-myristate 13-acetate, 97%
CAS: 16561-29-8 Molecular Formula: C36H56O8 Molecular Weight (g/mol): 616.84 MDL Number: MFCD00036736 InChI Key: PHEDXBVPIONUQT-RGYGYFBISA-N Synonym: phorbol 12-myristate 13-acetate,phorbol ester,12-o-tetradecanoylphorbol-13-acetate,12-o-tetradecanoylphorbol 13-acetate,factor a1,tetradecanoylphorbol acetate,12-tetradecanoylphorbol 13-acetate,factor a1 croton oil,phorbol 13-acetate 12-myristate,phorbol-12-myristate-13-acetate PubChem CID: 27924 ChEBI: CHEBI:37537 IUPAC Name: (1S,2S,6R,10S,11R,13S,14R,15R)-13-(acetyloxy)-1,6-dihydroxy-8-(hydroxymethyl)-4,12,12,15-tetramethyl-5-oxotetracyclo[8.5.0.0²,⁶.0¹¹,¹³]pentadeca-3,8-dien-14-yl tetradecanoate SMILES: CCCCCCCCCCCCCC(=O)O[C@@H]1[C@@H](C)[C@]2(O)[C@@H]3C=C(C)C(=O)[C@@]3(O)CC(CO)=C[C@H]2[C@@H]2C(C)(C)[C@]12OC(C)=O
Phorbol 12-myristate 13-acetate, 95%
CAS: 16561-29-8 Molecular Formula: C36H56O8 Molecular Weight (g/mol): 616.84 MDL Number: MFCD00036736 InChI Key: PHEDXBVPIONUQT-RGYGYFBISA-N Synonym: phorbol 12-myristate 13-acetate,phorbol ester,12-o-tetradecanoylphorbol-13-acetate,12-o-tetradecanoylphorbol 13-acetate,factor a1,tetradecanoylphorbol acetate,12-tetradecanoylphorbol 13-acetate,factor a1 croton oil,phorbol 13-acetate 12-myristate,phorbol-12-myristate-13-acetate PubChem CID: 27924 ChEBI: CHEBI:37537 IUPAC Name: (1S,2S,6R,10S,11R,13S,14R,15R)-13-(acetyloxy)-1,6-dihydroxy-8-(hydroxymethyl)-4,12,12,15-tetramethyl-5-oxotetracyclo[8.5.0.0²,⁶.0¹¹,¹³]pentadeca-3,8-dien-14-yl tetradecanoate SMILES: CCCCCCCCCCCCCC(=O)O[C@@H]1[C@@H](C)[C@]2(O)[C@@H]3C=C(C)C(=O)[C@@]3(O)CC(CO)=C[C@H]2[C@@H]2C(C)(C)[C@]12OC(C)=O
Phorbol 12-Myristate 13-acetate, Molecular biology reagent grade, MP Biomedicals™
CAS: 16561-29-8 Molecular Formula: C36H56O8 Molecular Weight (g/mol): 616.84 MDL Number: MFCD00036736 InChI Key: PHEDXBVPIONUQT-RGYGYFBISA-N Synonym: phorbol 12-myristate 13-acetate,phorbol ester,12-o-tetradecanoylphorbol-13-acetate,12-o-tetradecanoylphorbol 13-acetate,factor a1,tetradecanoylphorbol acetate,12-tetradecanoylphorbol 13-acetate,factor a1 croton oil,phorbol 13-acetate 12-myristate,phorbol-12-myristate-13-acetate PubChem CID: 27924 ChEBI: CHEBI:37537 IUPAC Name: (1S,2S,6R,10S,11R,13S,14R,15R)-13-(acetyloxy)-1,6-dihydroxy-8-(hydroxymethyl)-4,12,12,15-tetramethyl-5-oxotetracyclo[8.5.0.0²,⁶.0¹¹,¹³]pentadeca-3,8-dien-14-yl tetradecanoate SMILES: CCCCCCCCCCCCCC(=O)O[C@@H]1[C@@H](C)[C@]2(O)[C@@H]3C=C(C)C(=O)[C@@]3(O)CC(CO)=C[C@H]2[C@@H]2C(C)(C)[C@]12OC(C)=O
MilliporeSigma™ Calbiochem™ Phorbol-12-myristate-13-acetate
Phorbol-12-myristate-13-acetate, CAS 16561-29-8, is the most common phorbol ester. Activates PKC at nanomolar concentrations.
Phorbol 12-myristate 13-acetate, ≥99%, MP Biomedicals™
CAS: 16561-29-8 Molecular Formula: C36H56O8 Molecular Weight (g/mol): 616.84 MDL Number: MFCD00036736 InChI Key: PHEDXBVPIONUQT-RGYGYFBISA-N Synonym: phorbol 12-myristate 13-acetate,phorbol ester,12-o-tetradecanoylphorbol-13-acetate,12-o-tetradecanoylphorbol 13-acetate,factor a1,tetradecanoylphorbol acetate,12-tetradecanoylphorbol 13-acetate,factor a1 croton oil,phorbol 13-acetate 12-myristate,phorbol-12-myristate-13-acetate PubChem CID: 27924 ChEBI: CHEBI:37537 IUPAC Name: (1S,2S,6R,10S,11R,13S,14R,15R)-13-(acetyloxy)-1,6-dihydroxy-8-(hydroxymethyl)-4,12,12,15-tetramethyl-5-oxotetracyclo[8.5.0.0²,⁶.0¹¹,¹³]pentadeca-3,8-dien-14-yl tetradecanoate SMILES: CCCCCCCCCCCCCC(=O)O[C@@H]1[C@@H](C)[C@]2(O)[C@@H]3C=C(C)C(=O)[C@@]3(O)CC(CO)=C[C@H]2[C@@H]2C(C)(C)[C@]12OC(C)=O
MP Biomedicals™ Phorbol-12-myristate-13-acetate, >99%
Phorbol-12-myristate-13-acetate, >99% is a potent tumor promoter and activates protein kinase C in vivo and in vitro.
Phorbol 12-Myristate 13-acetate, ∽99%, Molecular biology reagent grade, MP Biomedicals™
CAS: 16561-29-8 Molecular Formula: C36H56O8 Molecular Weight (g/mol): 616.84 MDL Number: MFCD00036736 InChI Key: PHEDXBVPIONUQT-RGYGYFBISA-N Synonym: phorbol 12-myristate 13-acetate,phorbol ester,12-o-tetradecanoylphorbol-13-acetate,12-o-tetradecanoylphorbol 13-acetate,factor a1,tetradecanoylphorbol acetate,12-tetradecanoylphorbol 13-acetate,factor a1 croton oil,phorbol 13-acetate 12-myristate,phorbol-12-myristate-13-acetate PubChem CID: 27924 ChEBI: CHEBI:37537 IUPAC Name: (1S,2S,6R,10S,11R,13S,14R,15R)-13-(acetyloxy)-1,6-dihydroxy-8-(hydroxymethyl)-4,12,12,15-tetramethyl-5-oxotetracyclo[8.5.0.0²,⁶.0¹¹,¹³]pentadeca-3,8-dien-14-yl tetradecanoate SMILES: CCCCCCCCCCCCCC(=O)O[C@@H]1[C@@H](C)[C@]2(O)[C@@H]3C=C(C)C(=O)[C@@]3(O)CC(CO)=C[C@H]2[C@@H]2C(C)(C)[C@]12OC(C)=O
R&D Systems™ Phorbol 12-myristate 13-acetate
CAS: 16561-29-8 Molecular Formula: C36H56O8
Fisher Scientific Phorbol 12-Myristate 13-Acetate (Clear or Whitish Film), Fisher BioReagents™
CAS: 16561-29-8 Molecular Formula: C36H56O8 Molecular Weight (g/mol): 616.84 MDL Number: MFCD00036736 InChI Key: PHEDXBVPIONUQT-RGYGYFBISA-N Synonym: phorbol 12-myristate 13-acetate,phorbol ester,12-o-tetradecanoylphorbol-13-acetate,12-o-tetradecanoylphorbol 13-acetate,factor a1,tetradecanoylphorbol acetate,12-tetradecanoylphorbol 13-acetate,factor a1 croton oil,phorbol 13-acetate 12-myristate,phorbol-12-myristate-13-acetate PubChem CID: 27924 ChEBI: CHEBI:37537 IUPAC Name: (1S,2S,6R,10S,11R,13S,14R,15R)-13-(acetyloxy)-1,6-dihydroxy-8-(hydroxymethyl)-4,12,12,15-tetramethyl-5-oxotetracyclo[8.5.0.0²,⁶.0¹¹,¹³]pentadeca-3,8-dien-14-yl tetradecanoate SMILES: CCCCCCCCCCCCCC(=O)O[C@@H]1[C@@H](C)[C@]2(O)[C@@H]3C=C(C)C(=O)[C@@]3(O)CC(CO)=C[C@H]2[C@@H]2C(C)(C)[C@]12OC(C)=O