Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Phorbol-12-myristate-13-acetate, >99%

Catalog No. 0215186401 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-151-86401 1 mg
MP215186410 10 mg
MP215186425 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02-151-86401 Supplier MP Biomedicals™ Supplier No. 0215186401
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phorbol-12-myristate-13-acetate, >99% is a potent tumor promoter and activates protein kinase C in vivo and in vitro.

  • PMA′s effects on PKC are attributed to its similarity to diacylgylerol, a natural activator of PKC.
  • PMA activates Ca2+− ATPase and potentiates forskolin-induced cAMP formation.
  • It has been shown to inhibit apoptosis induced by the Fas antigen, but PMA induces apoptosis in HL-60 promyelocytic leukemia cells.
TRUSTED_SUSTAINABILITY

Specifications

Purity >98% (HPLC)
For Use With (Application) Potent tumor promoter, Activates protein kinase C in vivo and in vitro
Solubility Practically insoluble.
Molecular Formula C36H56O8
CAS 16561-29-8
Content And Storage Store at − 20°C. Protect from light.
Quantity 1 mg
Molecular Weight (g/mol) 616.84 g/mol
Melting Point 162°F
Vapor Pressure 1.8X10-17 mm Hg at 25°C
Form Solid
Show More Show Less

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.