Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Phorbol 12-Myristate 13-Acetate, PKC Activator, Solid, 25 mg

Catalog No. 502584631
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-4631 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-4631 Supplier Abcam Supplier No. AB12029725MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent nanomolar activator of protein kinase C with >99% purity, cited in over 45 publications.

  • Potency: Potent nanomolar activator of protein kinase C (PKC)
  • Mechanism: Binds to C1 domain of PKC, induces membrane translocation and enzyme activation
  • Additional Actions: Reported to act on non-kinase proteins including chimaerins, RasGRP, and Unc-13/Munc-13
  • Properties: Extremely potent mouse skin tumor promoter
  • Purity: > 99%
  • Citations: Cited in over 45 publications

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Protein Kinase C, Chimaerins, RasGRP, Unc-13/Munc-13
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Content And Storage Store at -20°C.
Form Solid
Formulation DMSO (10mM)
Quantity 25 mg
For Use With (Application) Functional studies, Immunocytochemistry/Immunofluorescence, Flow cytometry, Sandwich ELISA, Western blot, Immunoprecipitation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.