Learn More
Abcam Phorbol 12-myristate 13-acetate, PKC Activator, Solid, 5 mg
Description
- Potent Activator: Nanomolar activator of protein kinase C (PKC) in vivo and in vitro.
- Mechanism: Binds to the C1 domain of PKC, inducing membrane translocation and enzyme activation.
- Additional Actions: Reported to interact with non-kinase proteins including chimaerins, RasGRP, and Unc-13/Munc-13.
- Purity: > 99%.
- Physical State: Solid.
- Molecular Weight: 616.8 Da.
- Citations: Cited in over 45 publications.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Target Molecule | Protein Kinase C, Chimaerins, RasGRP, Unc-13/Munc-13 |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Quantity | 5 mg |
| For Use With (Application) | Functional studies, Immunocytochemistry/Immunofluorescence, Flow cytometry, Sandwich ELISA, Western blot, Immunoprecipitation |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.