Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Phorbol 12-myristate 13-acetate (PMA), PKC activator (DMSO solution), 1 mg

Catalog No. 502658067
Encompass_Preferred
Catalog No. 50-265-8067 Supplier Abcam Supplier No. AB1474651MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phorbol 12-myristate 13-acetate (PMA), activator.

  • Class: Activator
  • Form: Liquid
  • Purity: >99%
  • Molecular Weight: 616.8 g/mol

Specifications

Purity >99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.