Learn More
Cayman Chemical Phorbol 12-myristate 13-acetate, 16561-29-8, 1 mg
SDP
Supplier:  Cayman Chemical 100080141
Encompass_Preferred
Molecular Formula C36H56O8, Purity ≥95%, Formula Weight 616.8, Synonyms: PMA, 12-O-Tetradecanoylphorbol-13-acetate, TPA; A phorbol ester that is commonly used to activate certain types of protein PKC and, indirectly, certain MAP kinase pathways; prolonged treatment with PMA produces various effects, ranging from tumorigenesis to hematopoietic differentiation.
Catalog No. NC1522954