Learn More
Selleck Chemical LLC Phosphatase Inhibitor Cocktail B15001-1MLA 1MLB
SDP
Supplier:  Selleck Chemical LLC B150011MLA 1MLB
Encompass_Preferred
Phosphatase Inhibitor Cocktail for phosphorylation protection is provided in a 2 tube formula at 100X stock solution It combines the inhibitory capability of both inorganic salts and peptides
Catalog No. 50-000-39353