Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Phosphate, 0.2M buffer soln., pH 8.5

Catalog No. p-7076578
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61925AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ61925AP Supplier Thermo Scientific™ Supplier No. J61925AP

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Used in molecular biology, biochemistry, and chromatography. It is used in the purification of antibodies.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7558-80-7
Concentration 0.2M buffer soln.
Physical Form Liquid
Quantity 500 mL
Packaging Plastic Bottle
Molecular Weight (g/mol) 119.976
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.