Learn More
CPC Scientific H-Arg-Arg-Lys-Ala-Ser-Gly-Pro-Pro-Val-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific PKCP026A
Encompass
ONE-LETTER SEQUENCE: RRKASGPPVMOLECULAR FORMULA: C41H74N16O11MOLECULAR WEIGHT:967.15STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [93511-94-5], RESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES: C.A. O'Brian el al., BBRC, 124, 296 (1984)
Catalog No. 50-211-0176
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.