Learn More
Bioassay Systems QuantiChrom™ Phosphate Assay Kit. For quantitative determination of phosphate and evaluation of drug effects on phosphate metabolism.
SDP
Catalog No. 50489223 Shop All Bioassay Systems Products
Encompass_Preferred
For quantitative determination of phosphate and evaluation of drug effects on phosphate metabolism. Key Features: Sensitive and accurate. Linear detection range 0.3 µM (0.0028 mg/dL) to 50 µM (0.47 mg/dL) phosphate in 96-well plate assay. Simple and high-throughput. Can be readily automated as a high-throughput assay for thousands of samples per day. Improved reagent stability and versatility. The optimized formulation has greatly enhanced reagent and signal stability. Assays can be executed in cuvet or 96-well plate. Low interference in biological samples. No pretreatments are needed. Assays can be directly performed on raw biological samples i.e., in the presence of lipid, protein and minerals. Method: OD620nm (malachite green). Samples: Serum, urine, saliva, sweat, tissue culture, food, environment etc. Species: All. Procedure: Assay takes 30 min. Kit size: 500 tests. Detection limit: 3 µg/dL (0.3 µM).
Catalog No. 50-489-223 Supplier Bioassay Systems Supplier No. DIPI500
For quantitative determination of phosphate and evaluation of drug effects on phosphate metabolism. Key Features: Sensitive and accurate. Linear detection range 0.3 µM (0.0028 mg/dL) to 50 µM (0.47 mg/dL) phosphate in 96-well plate assay. Simple and high-throughput. Can be readily automated as a high-throughput assay for thousands of samples per day. Improved reagent stability and versatility. The optimized formulation has greatly enhanced reagent and signal stability. Assays can be executed in cuvet or 96-well plate. Low interference in biological samples. No pretreatments are needed. Assays can be directly performed on raw biological samples i.e., in the presence of lipid, protein and minerals. Method: OD620nm (malachite green). Samples: Serum, urine, saliva, sweat, tissue culture, food, environment etc. Species: All. Procedure: Assay takes 30 min. Kit size: 500 tests. Detection limit: 3 µg/dL (0.3 µM).