Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphate Buffer, 0.05 Molar, pH 7.4, Reagents

Catalog No. 01334156
Change view
Click to view available options
Quantity:
4 L
20 L
Packaging:
Cubitainer
HDPE Winpak
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-334-156 20 L HDPE Winpak
01-334-155 20 L Cubitainer
01-334-154 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-334-156 Supplier Reagents Holdings Llc Supplier No. CP20950020E7
Only null left
Add to Cart
Add to Cart

Phosphate Buffer, 0.05 Molar, pH 7.4, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Specifications

Chemical Name or Material Buffer
CAS 7732-18-5
Color Colorless
pH 7.4
Physical Form Liquid
Quantity 20 L
Packaging HDPE Winpak
Sterility Non-sterile
Solubility Information Completely miscible
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.