Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phosphate Buffer pH 6.2 to 6.5, For Chlorine, Chlorine Dioxide, Certified, LabChem™
$68.10 - $271.50
Specifications
pH Range | 6.2 to 6.5 |
---|---|
Format | Liquid |
Packaging | Poly Bottle |
Storage Requirements | Room Temperature |
Specifications
6.2 to 6.5 | |
Liquid | |
Poly Bottle | |
Room Temperature |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title