Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphate Buffer, pH 6.8, Reagents

Catalog No. 01334126
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-126 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-334-126 Supplier Reagents Holdings Llc Supplier No. CP23500020F
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phosphate Buffer, pH 6.8, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Buffer
CAS 7732-18-5
Color Colorless
pH 6.8
Physical Form Liquid
Quantity 20 L
Packaging Cubitainer
Sterility Non-sterile
Solubility Information Completely miscible
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.