Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphate Buffer Solution, pH 7, ASTM for Chlorine, Ricca Chemical
SDP

Catalog No. 58085 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
58085 20 L Cubitainer
5808-16 500 mL Natural Poly Bottle
R58080004A 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 58085 Supplier Ricca Chemical Company Supplier No. 58085
Add to Cart
Add to Cart

For Chlorine Determination (ASTM Formulation)

Specifications

Chemical Name or Material Phosphate Buffer Solution
Name Note pH 7
CAS 7732-18-5, 7558-79-4, 7778-77-0, 1310-73-2
CAS Min % 92.0, 3.14, 2.33, 0.34
Color Colorless
CAS Max % 96.0, 3.27, 2.43, 0.36
Concentration 1X
For Use With (Application) Water Analysis
pH 7
Physical Form Liquid
Quantity 20 L
Solution Type Buffer
Packaging Cubitainer
Sterility Non-sterile
Grade Laboratory
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.