Learn More
Bioassay Systems Malachite Green Phosphate Assay Kit. For sensitive and high-throughput phosphate determination.
SDP
Catalog No. 50489295 Shop All Bioassay Systems Products
Encompass_Preferred
For sensitive and high-throughput phosphate determination. Key Features: Reagent very stable. Due to our innovative formulation, no precipitation of reagent occurs. Therefore no filtration of reagent is needed prior to assays, as is often required with other commercial kits. High sensitivity and wide detection range: detection of as little of 1.6 pmoles of phosphate and useful range between 0.02 µM and 40 µM phosphate. Fast and convenient: homogeneous "mix-and-measure".Compatible with routine laboratory and HTS formats: assays can be performed in tubes, cuvettes or microplates, on spectrophotometers and plate readers. Robust and amenable to HTS: Z factors of 0.7 to 0.9 are observed in 96-well and 384-well plates. Can be readily automated on HTS liquid handling systems. Method: OD620nm (malachite green). Samples: Biological, environment etc. Species: All. Procedure: Assay takes 30 min. Kit size: 2500 tests. Detection limit: 0.02 µM.
Catalog No. 50-489-295 Supplier Bioassay Systems Supplier No. POMG25H
For sensitive and high-throughput phosphate determination. Key Features: Reagent very stable. Due to our innovative formulation, no precipitation of reagent occurs. Therefore no filtration of reagent is needed prior to assays, as is often required with other commercial kits. High sensitivity and wide detection range: detection of as little of 1.6 pmoles of phosphate and useful range between 0.02 µM and 40 µM phosphate. Fast and convenient: homogeneous "mix-and-measure".Compatible with routine laboratory and HTS formats: assays can be performed in tubes, cuvettes or microplates, on spectrophotometers and plate readers. Robust and amenable to HTS: Z factors of 0.7 to 0.9 are observed in 96-well and 384-well plates. Can be readily automated on HTS liquid handling systems. Method: OD620nm (malachite green). Samples: Biological, environment etc. Species: All. Procedure: Assay takes 30 min. Kit size: 2500 tests. Detection limit: 0.02 µM.