Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphatidyl-L-serine, ∽98%, MP Biomedicals™

Catalog No. p-4403866
Click to view available options
Quantity:
10 mg
50 mg

Chemical Identifiers

CAS 51446-62-9
Molecular Formula C42H82NO10P
Molecular Weight (g/mol) 792.089
InChI Key TZCPCKNHXULUIY-RGULYWFUSA-N
Synonym distearoyl phosphatidylserine, 2s-2-amino-3-r-2,3-bis stearoyloxy propoxy hydroxy phosphoryl oxy propanoic acid, phosphatidylserine 36:0, dioctadecanoylphosphatidylserine, pser 36:0, 1,2-dioctadecanoyl-sn-glycero-3-phosphoserine, phosphatidylserine 18:0/18:0, l-serine, 2,3-bis 1-oxooctadecyl oxy propyl ester, dihydrogen phosphate ester , r, 1,2-distearoyl-sn-glycero-3-phospho-l-serine, l-alpha-phosphatidyl-l-serine
PubChem CID 9547096
ChEBI CHEBI:84519
IUPAC Name (2S)-2-amino-3-[[(2R)-2,3-di(octadecanoyloxy)propoxy]-hydroxyphosphoryl]oxypropanoic acid
SMILES CCCCCCCCCCCCCCCCCC(=O)OCC(COP(=O)(O)OCC(C(=O)O)N)OC(=O)CCCCCCCCCCCCCCCCC

Specifications

Color Yellow
Assay Percent Range ∽ 98%
Quantity 10 mg
Formula Weight 788
Physical Form Liquid
Chemical Name or Material Phosphatidyl-L-serine
Recommended Storage Store at 0°C.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.