Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphatidylethanol, 98%, MP Biomedicals™

Catalog No. ICN15898350 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15898350 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15898350 Supplier MP Biomedicals, Inc Supplier No. 0215898350

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

Molecular Formula C39H75O8P
Synonym 1-Palmitoyl-2-oleoyl-sn-glycero-3-phosphoethanol

Specifications

Assay Percent Range 98%
Quantity 50 mg
Formula Weight 703
Physical Form Solid
Chemical Name or Material Phosphatidylethanol
Recommended Storage Store at -20°C.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.