Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-4EBP1 (Thr37, Thr46) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA577963
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA577963 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA577963 Supplier Invitrogen™ Supplier No. PA577963
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: Neuro2A, C8D30, NIH-3T3, Raw264.7, C2C12, PC-12, Rat2, HeLa, 293T (starvation for 24 hr), 293T (starvation for 24 hr and 20%FBS treatment for 0.5 hr) Predicted Reactivity: Zebrafish (100%), Pig (100%), Bovine (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes one member of a family of translation repressor proteins. The protein directly interacts with eukaryotic translation initiation factor 4E, which is a limiting component of the multisubunit complex that recruits 40S ribosomal subunits to the 5' end of mRNAs. Interaction of this protein with eIF4E inhibits complex assembly and represses translation. This protein is phosphorylated in response to various signals including UV irradiation and insulin signaling, resulting in its dissociation from eIF4E and activation of mRNA translation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-4EBP1 (Thr37, Thr46)
Applications Western Blot
Classification Polyclonal
Concentration 0.12 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene EIF4EBP1
Gene Accession No. Q13541, Q60876, Q62622
Gene Alias 4EBP1; 4E-BP1; AA959816; BP-1; eIF4E-binding protein 1; EIF4EBP1; eukaryotic translation initiation factor 4E binding protein 1; eukaryotic translation initiation factor 4E-binding protein 1; MGC431; MGC4316; P/OKCL.6; PHAS-1; PHAS-I; Phosphorylated heat- and acid-stable protein regulated by insulin 1
Gene Symbols EIF4EBP1
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide surrounding phospho Thr37/Thr46 of human 4E-BP1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116636, 13685, 1978
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.