Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-53BP1 (Ser1778) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA517462 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA517462 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA517462 Supplier Invitrogen™ Supplier No. PA517462
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

It is not recommended to aliquot this antibody.

53BP1 (P53-binding protein 1) plays a critical role in tumor suppression and is a putative substrate of ATM kinase. Upon DNA damage, 53BP1 is phosphorylated and localizes to the presumptive sites of damage, specifically, double-strand breaks. 53BP1 may have a role in checkpoint signaling during mitosis, enhances TP53-mediated transcriptional activation, and participates in DNA repair by maintaining genomic stability.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-53BP1 (Ser1778)
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 32 μg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5
Gene TP53BP1
Gene Accession No. Q12888
Gene Alias 53BP1; FLJ41424; m53BP1; MGC138366; murine p53-binding protein; p202; p53-binding protein 1; p53BP1; TDRD30; TP53; TP53-binding protein 1; Tp53bp1; transformation related protein 53 binding protein 1; transformation related protein 53 binding-protein 1; Trp53bp1; tumor protein 53-binding protein, 1; tumor protein p53 binding protein 1; tumor protein p53-binding protein, 1; tumor suppressor p53-binding protein 1
Gene Symbols TP53BP1
Host Species Rabbit
Immunogen Synthetic phosphopeptide corresponding to residues surrounding pSer1778 of human 53BP1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7158
Target Species Human, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.