Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho alpha-Synuclein (S129) Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. PPS091
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PPS091 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PPS091 Supplier R&D Systems Supplier No. PPS091
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

alpha-Synuclein Polyclonal specifically detects alpha-Synuclein in Human, Mouse, Rat, Bovine, Canine, Primate samples. It is validated for Western Blot.

Specifications

Antigen alpha-Synuclein (p ser129)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000 dilution
Gene Alias alpha-synuclein, I+/--synuclein, MGC110988, NACP, non A-beta component of AD amyloid, Non-A beta component of AD amyloid, Non-A4 component of amyloid precursor, PARK1, PARK4, PD1, SNCA, synuclein alpha-140, synuclein, alpha (non A4 component of amyloid precursor), Synuclein-alpha, truncated alpha synuclein
Gene Symbols SNCA
Host Species Rabbit
Immunogen Phosphopeptide corresponding to amino acid residues surrounding the phospho-S129 of Synuclein-alpha
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6622
Test Specificity Human, mouse, rat, bovine, canine, and non-human primate Synuclein-alpha phosphorylated at S129
Target Species Human, Mouse, Rat, Bovine, Canine, Primate
Content And Storage For long-term storage, < -20degrees C is recommended. Product is stable at < -20degrees C for at least 1 year.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.