Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-AMPK alpha-2 (Thr172) Recombinant Rabbit Monoclonal Antibody (4F4)
GREENER_CHOICE

Catalog No. PIMA533216 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533216 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533216 Supplier Invitrogen™ Supplier No. MA533216
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

Phospho-AMPK alpha-2 (Thr172) Recombinant Monoclonal Antibody for Western Blot, ICC/IF, ELISA

The protein encoded by this gene is a catalytic subunit of the AMP-activated protein kinase (AMPK). AMPK is a heterotrimer consisting of an alpha catalytic subunit, and non-catalytic beta and gamma subunits. AMPK is an important energy-sensing enzyme that monitors cellular energy status. In response to cellular metabolic stresses, AMPK is activated, and thus phosphorylates and inactivates acetyl-CoA carboxylase (ACC) and beta-hydroxy beta-methylglutaryl-CoA reductase (HMGCR), key enzymes involved in regulating de novo biosynthesis of fatty acid and cholesterol. Studies of the mouse counterpart suggest that this catalytic subunit may control whole-body insulin sensitivity and is necessary for maintaining myocardial energy homeostasis during ischemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-AMPK alpha-2 (Thr172)
Applications ELISA, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 4F4
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PRKAA2
Gene Accession No. P54646, Q8BRK8
Gene Alias 2310008I11Rik; 5'-AMP-activated protein kinase catalytic subunit alpha-2; 5'-AMP-activated protein kinase, catalytic alpha-2 chain; A830082D05; ACACA kinase; Acetyl-CoA carboxylase kinase; AMP-activated protein kinase; AMP-activated protein kinase alpha-2 subunit variant 2; AMP-activated protein kinase alpha-2 subunit variant 3; AMP-activated protein kinase alpha-2 variant B; AMPK; AMPK alpha 2; AMPK alpha-2 chain; AMPK subunit alpha-2; AMPK2; AMPKa2; AMPKalpha2; AMPK-alpha-2 chain; EC 2.7.11.1; HMGCR kinase; hydroxymethylglutaryl-CoA reductase kinase; PRKAA; Prkaa2; protein kinase AMP-activated catalytic subunit alpha 2; protein kinase, AMP-activated, alpha 2 catalytic subunit
Gene Symbols PRKAA2
Host Species Rabbit
Immunogen A synthesized peptide derived from human Phospho-PRKAA2 (Thr172).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108079, 5563
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.