Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-ATF4 (Ser245) Polyclonal Antibody, QED Bioscience™
SDP

Catalog No. 502278087
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-8087 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-8087 Supplier Leinco Technologies Inc Supplier No. 43053100UL
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Phospho-ATF4 (Ser245) Polyclonal antibody specifically detects Phospho-ATF4 (Ser245) in Human samples. It is validated for Immunohistochemistry, Western Blot

Specifications

Antigen Phospho-ATF4 (Ser245)
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Dulbecco's PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Atf4
Gene Accession No. P18848
Gene Alias Activating transcription factor 4; activating transcription factor 4 (tax-responsive enhancer element B67); activating transcription factor 4a; activating transcription factor 4b1 (tax-responsive enhancer element B67); activating transcription factor ATF-4; ATF 4; atf4; Atf-4; atf4 {ECO:0000312; ATF4 protein; atf4a; atf4b1; C/ATF; C/EBP-related ATF; cAMP response element-binding protein 2; cAMP-dependent transcription factor ATF-4; cAMP-responsive element-binding protein 2; CREB 2; CREB2; CREB-2; Cyclic AMP-dependent transcription factor ATF-4; cyclic AMP-responsive element-binding protein 2; DNA-binding protein TAXREB67; hypothetical protein LOC509107; OTTHUMP00000199130; rATF-4; RGD:621863}; tax-re; TaxREB67; TaxREB67 homolog; tax-responsive enhancer element B67; tax-responsive enhancer element-binding protein 67; Tax-responsive enhancer element-binding protein 67 homolog; TXREB; wu:fj01c08; wu:fk30b04; zgc:85951
Host Species Rabbit
Immunogen Peptide sequence that includes phosphorylation sites of Serine 245 (N-R-S(p)-L-P) derived from human ATF4 and conjugated to KLH.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 468
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.