Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-Aurora A (Thr288) Polyclonal Antibody
GREENER_CHOICE

Catalog No. 441210G
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-121-0G 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-121-0G Supplier Invitrogen™ Supplier No. 441210G
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

Phospho-Aurora A (Thr288) Polyclonal Antibody for ICC/IF, IHC (P), Western Blot

AURKA (Aurora A) is a serine/threonine kinase that regulates mitosis by association in vivo with centrosomes. Overexpression of AURKA is observed in many types of cancer. Aurora A plays a role in cell cycle regulation during anaphase and/or telophase, in relation to the function of the centrosome/spindle pole region during chromosome segregation. Aurora A plays a key role during tumor development and progression and is overexpressed in many human cancers including breast, ovarian and colorectal. Aurora A is viewed as a potential target for anticancer drug treatment. Tissue specificity: Highly expressed in testis and weakly in skeletal muscle, thymus and spleen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Aurora A (Thr288)
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation Dulbecco's PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.3
Gene AURKA
Gene Accession No. O14965, P97477
Gene Alias Aik; Airk; AIRK1; Ark1; ARK-1; AU019385; AURA; AURAK; AURKA; Aurora 2; aurora A; Aurora A Kinase; aurora family kinase 1; Aurora kinase A; aurora/IP; aurora/IPL1-like kinase; aurora/IPL1-related kinase 1; AURORA2; Aurora-A; aurora-related kinase 1; AW539821; Ayk1; Breast tumor-amplified kinase; BTAK; EC 2.7.11.1; hARK1; IAK; IAK1; Ipl1- and aurora-related kinase 1; IPL1-related kinase; MGC34538; OTTHUMP00000031341; OTTHUMP00000166072; PPP1R47; protein phosphatase 1, regulatory subunit 47; ratAurA; RP5-1167H4.6; sereine threonine kinase; serine/threonine kinase 6; serine/threonine protein kinase 15; serine/threonine protein kinase 6; serine/threonine-protein kinase 15; Serine/threonine-protein kinase 6; serine/threonine-protein kinase 6; aurora kinase A; serine/threonine-protein kinase aurora-A; Serine/threonine-protein kinase Ayk1; STK15; STK6; STK7; Unknown (protein for MGC:134472)
Gene Symbols AURKA
Host Species Rabbit
Immunogen The antiserum was produced against a chemically synthesized phosphopeptide derived from the region of human Aurora A Kinase that contains threonine 288. The sequence is highly conserved in mouse and rat.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20878, 6790
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.