Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-Aurora B/C (Thr236, Thr202) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA536030
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA536030 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA536030 Supplier Invitrogen™ Supplier No. PA536030
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 35 and 39 kDa. Purity is >95% by SDS-PAGE.

Aurora kinases belong to a highly conserved family of mitotic serine/threonine kinases with three members identified among mammals: Aurora A, B, and C. Studies on the temporal expression pattern and subcellular localization of Aurora kinases in mitotic cells suggest an association with mitotic structure. Aurora kinase functional influences span from G2 phase to cytokinesis and may be involved in key cell cycle events such as centrosome duplication, chromosome bi-orientation and segregation, cleavage furrow positioning, and ingression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Aurora B/C (Thr236, Thr202)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.2
Gene AURKC
Gene Accession No. O55099, O70126, O88445, Q96GD4, Q9UQB9
Gene Alias AIE1; AIE2; Aik2; AIK3; Aim1; AIM-1; AIRK2; AIRK3; AL022959; ARK2; ARK-2; ARK3; ARK-3; AurB; AurC; Aurkb; aurkb-sv1; aurkb-sv2; Aurkc; Aurora 1; aurora 3; Aurora- and IPL1-like midbody-associated protein 1; aurora B; aurora kinase B; aurora kinase B-Sv1; aurora kinase B-Sv2; aurora kinase C; aurora/Ipl1/Eg2 protein 1; aurora/IPL1/EG2 protein 2; aurora/IPL1-related kinase 2; Aurora/IPL1-related kinase 3; aurora-1; aurora-B; aurora-C; aurora-related kinase 2; aurora-related kinase 3; epididymis secretory protein Li 90; HEL-S-90; IAK3; IPL1; PPP1R48; protein phosphatase 1, regulatory subunit 48; serine/threonine kinase 12; serine/threonine kinase 13 (aurora/IPL1-like); serine/threonine kinase 13 (aurora/IPL-like); serine/threonine kinase 5; serine/threonine-protein kinase 12; Serine/threonine-protein kinase 13; serine/threonine-protein kinase 5; Serine/threonine-protein kinase aurora-B; serine/threonine-protein kinase aurora-C; SPGF5; Stk1; STK-1; STK12; Stk13; Stk5
Gene Symbols AURKB, AURKC
Host Species Rabbit
Immunogen Synthetic phosphopeptide derived from human AURKB/C around the phosphorylation site of Threonine 236/202.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114592, 20871, 20877, 292554, 6795, 9212
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.